siRNA Details
| Gene Symbol: Nmral1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191588.1 | NP_001178517.1 | 4 | 364 | ||
| XM_006245784.1 | XP_006245846.1 | 3 | 643 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nmral1| Gene Symbol: Nmral1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191588.1 | NP_001178517.1 | 4 | 364 | ||
| XM_006245784.1 | XP_006245846.1 | 3 | 643 | |||