siRNA Details
| Gene Symbol: Olr1376 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_214827.1 | NP_999992.1 | 1 | 306 | ||
| Gene Symbol: Olr1382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_214831.1 | NP_999996.1 | 1 | 306 | ||

Olr1376
Genomic Map