siRNA Details
| Gene Symbol: Prss30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199371.1 | NP_955403.1 | 4 | 389 | ||
| XM_006245922.2 | XP_006245984.1 | 5 | 580 | |||
| XM_006245923.2 | XP_006245985.1 | 5 | 588 | |||
| XM_008767553.1 | XP_008765775.1 | 5 | 608 | |||
| GenBank mRNA | AB073023.1 | - | 4 | 389 | ||

Prss30
Genomic Map