siRNA Details
| Gene Symbol: Prss27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182949.3 | NP_891994.3 | 3, 4 | 320 | ||
| GenBank mRNA | AB101677.1 | - | 3, 4 | 320 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prss27| Gene Symbol: Prss27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182949.3 | NP_891994.3 | 3, 4 | 320 | ||
| GenBank mRNA | AB101677.1 | - | 3, 4 | 320 | ||