siRNA Details
| Gene Symbol: Rnps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011890.1 | NP_001011890.1 | 3 | 313 | ||
| XM_006245931.2 | XP_006245993.1 | 3 | 757 | |||
| XM_006245932.2 | XP_006245994.1 | 4 | 456 | |||
| XM_006245933.2 | XP_006245995.1 | 2 | 252 | |||
| GenBank mRNA | BC079014.1 | - | 3 | 313 | 7113485 | |

Rnps1
Genomic Map