siRNA Details
Gene Symbol: Zfp598 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105770.1 | NP_001099240.1 | 11 | 2309 | ||
XM_006245934.2 | XP_006245996.1 | 11 | 2357 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp598 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105770.1 | NP_001099240.1 | 11 | 2309 | ||
XM_006245934.2 | XP_006245996.1 | 11 | 2357 |