siRNA Details
| Gene Symbol: Tbl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008277.1 | NP_001008278.1 | 7 | 673 | ||
| GenBank mRNA | BC085837.1 | - | 7 | 673 | 7135552 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbl3| Gene Symbol: Tbl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008277.1 | NP_001008278.1 | 7 | 673 | ||
| GenBank mRNA | BC085837.1 | - | 7 | 673 | 7135552 | |
Set Membership: |
|