siRNA Details
| Gene Symbol: Nubp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011891.1 | NP_001011891.1 | 3, 4 | 359 | ||
| XM_008767558.1 | XP_008765780.1 | 3, 4 | 372 | |||
| GenBank mRNA | BC079386.1 | - | 3, 4 | 359 | 7111553 | |
| FQ213066.1 | - | 3, 4 | 373 | |||

Nubp2
Genomic Map