siRNA Details
| Gene Symbol: Rpusd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105774.1 | NP_001099244.1 | 4 | 525 | ||
| XM_006245937.2 | XP_006245999.1 | 4 | 580 | |||
| GenBank mRNA | BC161966.1 | - | 4 | 523 | 9028415 | |
| FQ213277.1 | - | 4 | 446 | |||

Rpusd1
Genomic Map