siRNA Details
| Gene Symbol: Stub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025625.2 | NP_001020796.2 | 3 | 489 | ||
| GenBank mRNA | BC099240.1 | - | 3 | 436 | 7445727 | |
| CB568781.1 | - | 3 | 490 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stub1| Gene Symbol: Stub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025625.2 | NP_001020796.2 | 3 | 489 | ||
| GenBank mRNA | BC099240.1 | - | 3 | 436 | 7445727 | |
| CB568781.1 | - | 3 | 490 | |||