siRNA Details
| Gene Symbol: LOC102555298 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006246066.2 | XP_006246128.1 | 8 | 1307 | ||
| XM_006246068.2 | XP_006246130.1 | 8 | 1307 | |||
| XM_006246069.2 | XP_006246131.1 | 5 | 548 | |||
| XM_008767618.1 | XP_008765840.1 | 8 | 1361 | |||

LOC102555298
Genomic Map