siRNA Details
| Gene Symbol: Foxi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105776.1 | NP_001099246.1 | 2 | 910 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Foxi1| Gene Symbol: Foxi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105776.1 | NP_001099246.1 | 2 | 910 | ||