siRNA Details
| Gene Symbol: RGD1559575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_220362.6 | XP_220362.3 | 2 | 1797 | ||
| GenBank mRNA | BC158643.1 | - | 2 | 1500 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1559575| Gene Symbol: RGD1559575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_220362.6 | XP_220362.3 | 2 | 1797 | ||
| GenBank mRNA | BC158643.1 | - | 2 | 1500 | ||