siRNA Details
| Gene Symbol: Olr1385 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_214833.1 | NP_999998.1 | 1 | 659 | ||
| Gene Symbol: Olr1383 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_214832.1 | NP_999997.1 | 1 | 659 | ||
| Gene Symbol: Olr1384 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002291.1 | NP_001002291.1 | 1 | 653 | ||

Olr1385
Genomic Map