siRNA Details
| Gene Symbol: Olr1387 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000001.1 | NP_001000001.1 | 1 | 673 | ||
| Gene Symbol: Olr1388 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000788.1 | NP_001000788.1 | 1 | 673 | ||

Olr1387
Genomic Map