siRNA Details
| Gene Symbol: TRAPPC10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003274.4 | NP_003265.3 | 4 | 608 | ||
| XM_005261168.4 | XP_005261225.1 | 4 | 601 | |||
| XM_011529714.2 | XP_011528016.1 | 4 | 601 | |||
| XM_011529715.2 | XP_011528017.1 | 4 | 601 | |||
| XM_011529716.2 | XP_011528018.1 | 4 | 601 | |||
| XM_011529717.2 | XP_011528019.1 | 4 | 601 | |||
| XM_011529718.2 | XP_011528020.1 | 4 | 601 | |||
| XM_011529719.2 | XP_011528021.1 | 4 | 601 | |||
| XM_011529720.2 | XP_011528022.1 | 4 | 601 | |||
| XM_011529721.2 | XP_011528023.1 | 1 | 87 | |||
| XM_011529723.2 | XP_011528025.1 | 1 | 33 | |||
| XM_011529725.2 | XP_011528027.1 | 4 | 601 | |||
| XM_011529726.2 | XP_011528028.1 | 4 | 601 | |||
| XM_011529728.2 | XP_011528030.1 | 1 | 118 | |||
| XM_011529730.2 | XP_011528032.1 | 4 | 601 | |||
| XM_017028454.1 | XP_016883943.1 | 4 | 601 | |||
| GenBank mRNA | AL559301.3 | - | 4 | 614 | ||
| BC046241.1 | - | 4 | 586 | |||
| BC052247.1 | - | 4 | 586 | |||
| BC094823.1 | - | 4 | 586 | |||
| BC101728.1 | - | 4 | 528 | 8069234 | ||
| BX370317.2 | - | 1 | 88 | |||
| U19252.1 | - | 4 | 576 | |||
| U61500.1 | - | 4 | 592 | |||

TRAPPC10
Genomic Map