siRNA Details
| Gene Symbol: Sar1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009622.1 | NP_001009622.1 | 2, 3 | 145 | ||
| GenBank mRNA | BC088842.1 | - | 2, 3 | 145 | 7303357 | |
| FQ217751.1 | - | 2, 3 | 206 | |||
| FQ219381.1 | - | 2, 3 | 206 | |||

Sar1b
Genomic Map