siRNA Details
| Gene Symbol: Sar1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009622.1 | NP_001009622.1 | 5 | 344 | ||
| GenBank mRNA | BC088842.1 | - | 5 | 344 | 7303357 | |
| FQ215352.1 | - | 3 | 179 | |||
| FQ215892.1 | - | 3 | 179 | |||
| FQ216359.1 | - | 3 | 179 | |||
| FQ216811.1 | - | 3 | 179 | |||
| FQ217315.1 | - | 3 | 179 | |||
| FQ217751.1 | - | 5 | 405 | |||
| FQ219007.1 | - | 3 | 179 | |||
| FQ219108.1 | - | 3 | 179 | |||
| FQ219381.1 | - | 5 | 406 | |||
| FQ220447.1 | - | 3 | 179 | |||
| FQ224969.1 | - | 3 | 179 | |||
| FQ227071.1 | - | 3 | 179 | |||
| FQ233866.1 | - | 3 | 179 | |||

Sar1b
Genomic Map