siRNA Details
| Gene Symbol: Cdkn2aipnl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008278.1 | NP_001008279.1 | 1, 2 | 245 | ||
| GenBank mRNA | BC086396.1 | - | 1, 2 | 245 | 7303530 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdkn2aipnl| Gene Symbol: Cdkn2aipnl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008278.1 | NP_001008279.1 | 1, 2 | 245 | ||
| GenBank mRNA | BC086396.1 | - | 1, 2 | 245 | 7303530 | |