siRNA Details
| Gene Symbol: RGD1308564 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006246412.2 | XP_006246474.1 | 7 | 1229 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1308564| Gene Symbol: RGD1308564 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006246412.2 | XP_006246474.1 | 7 | 1229 | ||