siRNA Details
| Gene Symbol: Olr1425 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000010.1 | NP_001000010.1 | 1 | 786 | ||
| Gene Symbol: Olr1424 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000779.1 | NP_001000779.1 | 1 | 783 | ||

Olr1425
Genomic Map