siRNA Details
| Gene Symbol: Olr1428 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000011.1 | NP_001000011.1 | 1 | 60 | ||
| Gene Symbol: Olr1424 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000779.1 | NP_001000779.1 | 1 | 57 | ||

Olr1428
Genomic Map