siRNA Details
| Gene Symbol: Obscn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767886.1 | XP_008766108.1 | 109 | 24809 | ||
| GenBank mRNA | FQ224413.1 | - | 1 | 133 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Obscn| Gene Symbol: Obscn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767886.1 | XP_008766108.1 | 109 | 24809 | ||
| GenBank mRNA | FQ224413.1 | - | 1 | 133 | ||