siRNA Details
| Gene Symbol: Shmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047842.1 | NP_001041307.1 | 13 | 1662 | ||
| XM_006246469.2 | XP_006246531.1 | 10 | 1445 | |||
| GenBank mRNA | AY383687.1 | - | 13 | 1662 | ||
| BC099219.1 | - | 6 | 676 | |||

Shmt1
Genomic Map