siRNA Details
| Gene Symbol: B9d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105786.1 | NP_001099256.1 | 5, 6 | 455 | ||
| XM_006246553.2 | XP_006246615.1 | 1161 | ||||
| Gene Symbol: LOC100911746 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750843.3 | XP_003750891.1 | 5, 6 | 531 | ||
| XM_006246547.2 | XP_006246609.1 | 5, 6 | 1161 | |||

B9d1
Genomic Map