siRNA Details
| Gene Symbol: Map2k4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030023.1 | NP_001025194.1 | 4 | 554 | ||
| GenBank mRNA | AY880882.1 | - | 4 | 554 | ||
| KC795688.1 | - | 5 | 587 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Map2k4| Gene Symbol: Map2k4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030023.1 | NP_001025194.1 | 4 | 554 | ||
| GenBank mRNA | AY880882.1 | - | 4 | 554 | ||
| KC795688.1 | - | 5 | 587 | |||