siRNA Details
| Gene Symbol: Tmem220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_213341.8 | XP_213341.7 | 1, 2 | 160 | ||
| GenBank mRNA | FQ219853.1 | - | 1, 2 | 98 | ||
| FQ227969.1 | - | 1, 2 | 107 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem220| Gene Symbol: Tmem220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_213341.8 | XP_213341.7 | 1, 2 | 160 | ||
| GenBank mRNA | FQ219853.1 | - | 1, 2 | 98 | ||
| FQ227969.1 | - | 1, 2 | 107 | |||