siRNA Details
| Gene Symbol: Arhgef15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105789.1 | NP_001099259.1 | 5, 6 | 1151 | ||
| XM_008767788.1 | XP_008766010.1 | 5, 6 | 2445 | |||
| XM_008767789.1 | XP_008766011.1 | 5, 6 | 3005 | |||
| XM_008767790.1 | XP_008766012.1 | 4, 5 | 1334 | |||
| XM_008767791.1 | XP_008766013.1 | 4, 5 | 1335 | |||
| XM_008767792.1 | XP_008766014.1 | 4, 5 | 1333 | |||
| XM_008767793.1 | XP_008766015.1 | 4, 5 | 1336 | |||

Arhgef15
Genomic Map