siRNA Details
| Gene Symbol: Arhgef15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105789.1 | NP_001099259.1 | 9, 10 | 1796 | ||
| XM_008767788.1 | XP_008766010.1 | 8, 9 | 3243 | |||
| XM_008767789.1 | XP_008766011.1 | 8, 9 | 3803 | |||
| XM_008767790.1 | XP_008766012.1 | 7, 8 | 2132 | |||
| XM_008767791.1 | XP_008766013.1 | 8, 9 | 1980 | |||
| XM_008767792.1 | XP_008766014.1 | 8, 9 | 1978 | |||
| XM_008767793.1 | XP_008766015.1 | 7, 8 | 2134 | |||

Arhgef15
Genomic Map