siRNA Details
| Gene Symbol: Rangrf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105790.1 | NP_001099260.1 | 3 | 396 | ||
| GenBank mRNA | FQ228156.1 | - | 3 | 398 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rangrf| Gene Symbol: Rangrf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105790.1 | NP_001099260.1 | 3 | 396 | ||
| GenBank mRNA | FQ228156.1 | - | 3 | 398 | ||