siRNA Details
| Gene Symbol: Per1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034125.1 | NP_001029297.1 | 1 | 266 | ||
| XM_006246613.2 | XP_006246675.1 | 2 | 513 | |||
| XM_006246614.2 | XP_006246676.1 | 2 | 4163 | |||
| GenBank mRNA | AB092976.1 | - | 1 | 209 | ||
| AY903228.1 | - | 1 | 266 | |||
| AY903229.1 | - | 1 | 266 | |||
| AY903230.1 | - | 1 | 275 | |||

Per1
Genomic Map