siRNA Details
| Gene Symbol: Per1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034125.1 | NP_001029297.1 | 5 | 754 | ||
| XM_006246613.2 | XP_006246675.1 | 6 | 1001 | |||
| XM_006246614.2 | XP_006246676.1 | 6 | 4651 | |||
| GenBank mRNA | AB092976.1 | - | 5 | 697 | ||
| AY903228.1 | - | 5 | 754 | |||
| AY903229.1 | - | 5 | 754 | |||
| AY903230.1 | - | 5 | 763 | |||

Per1
Genomic Map