siRNA Details
| Gene Symbol: Aloxe3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105793.1 | NP_001099263.1 | 1 | 139 | ||
| XM_006246617.2 | XP_006246679.1 | 2 | 771 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aloxe3| Gene Symbol: Aloxe3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105793.1 | NP_001099263.1 | 1 | 139 | ||
| XM_006246617.2 | XP_006246679.1 | 2 | 771 | |||