siRNA Details
| Gene Symbol: Wrap53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007610.1 | NP_001007611.1 | 8 | 1347 | ||
| GenBank mRNA | BC083696.1 | - | 8 | 1347 | 7189640 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wrap53| Gene Symbol: Wrap53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007610.1 | NP_001007611.1 | 8 | 1347 | ||
| GenBank mRNA | BC083696.1 | - | 8 | 1347 | 7189640 | |