siRNA Details
| Gene Symbol: Zbtb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006246843.2 | XP_006246905.1 | 4 | 1310 | ||
| XM_006246844.2 | XP_006246906.1 | 3 | 1134 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zbtb4| Gene Symbol: Zbtb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006246843.2 | XP_006246905.1 | 4 | 1310 | ||
| XM_006246844.2 | XP_006246906.1 | 3 | 1134 | |||