siRNA Details
| Gene Symbol: Ctdnep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100494.1 | NP_001093964.1 | 3 | 410 | ||
| XM_006246643.2 | XP_006246705.1 | 4 | 535 | |||
| XM_006246644.2 | XP_006246706.1 | 4 | 535 | |||
| XM_008767804.1 | XP_008766026.1 | 3 | 439 | |||
| GenBank mRNA | BC087638.1 | - | 1 | 44 | ||
| BC107474.1 | - | 2 | 71 | |||
| BU671232.1 | - | 3 | 410 | |||

Ctdnep1
Genomic Map