siRNA Details
| Gene Symbol: Slc16a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005530.1 | NP_001005530.1 | 3 | 1267 | ||
| GenBank mRNA | BC081902.1 | - | 3 | 1267 | 7110851 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc16a13| Gene Symbol: Slc16a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005530.1 | NP_001005530.1 | 3 | 1267 | ||
| GenBank mRNA | BC081902.1 | - | 3 | 1267 | 7110851 | |