siRNA Details
| Gene Symbol: RGD1308134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127521.2 | NP_001120993.1 | 3 | 210 | ||
| XM_006246652.2 | XP_006246714.1 | 3 | 378 | |||
| XM_006246654.2 | XP_006246716.1 | 3 | 378 | |||
| GenBank mRNA | BC158879.1 | - | 3 | 210 | 7936013 | |

RGD1308134
Genomic Map