siRNA Details
| Gene Symbol: Med11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105799.1 | NP_001099269.1 | 2 | 258 | ||
| XM_006246660.2 | XP_006246722.1 | 2 | 596 | |||
| GenBank mRNA | FQ221563.1 | - | 2 | 270 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Med11| Gene Symbol: Med11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105799.1 | NP_001099269.1 | 2 | 258 | ||
| XM_006246660.2 | XP_006246722.1 | 2 | 596 | |||
| GenBank mRNA | FQ221563.1 | - | 2 | 270 | ||