siRNA Details
Gene Symbol: Wscd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024234.1 | NP_001019405.1 | 6 | 1558 | ||
GenBank mRNA | BC094520.1 | - | 6 | 1558 | 7316275 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Wscd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024234.1 | NP_001019405.1 | 6 | 1558 | ||
GenBank mRNA | BC094520.1 | - | 6 | 1558 | 7316275 |