siRNA Details
| Gene Symbol: Nek8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105804.1 | NP_001099274.1 | 5 | 724 | ||
| XM_006246933.2 | XP_006246995.1 | 5 | 2640 | |||
| XM_008767958.1 | XP_008766180.1 | 5 | 2640 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nek8| Gene Symbol: Nek8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105804.1 | NP_001099274.1 | 5 | 724 | ||
| XM_006246933.2 | XP_006246995.1 | 5 | 2640 | |||
| XM_008767958.1 | XP_008766180.1 | 5 | 2640 | |||