siRNA Details
| Gene Symbol: Shpk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033682.1 | NP_001028854.1 | 2 | 205 | ||
| GenBank mRNA | BC101914.1 | - | 2 | 205 | 7935768 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Shpk| Gene Symbol: Shpk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033682.1 | NP_001028854.1 | 2 | 205 | ||
| GenBank mRNA | BC101914.1 | - | 2 | 205 | 7935768 | |