siRNA Details
| Gene Symbol: Olr1466 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000024.1 | NP_001000024.1 | 1 | 622 | ||
| XM_008767806.1 | XP_008766028.1 | 2 | 1940 | |||
| Gene Symbol: Olr1467 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000025.1 | NP_001000025.1 | 1 | 631 | ||

Olr1466
Genomic Map