siRNA Details
| Gene Symbol: Olr1512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001350.1 | NP_001001350.1 | 1 | 850 | ||
| Gene Symbol: Olr1511 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000034.1 | NP_001000034.1 | 1 | 850 | ||

Olr1512
Genomic Map