siRNA Details
| Gene Symbol: Olr1513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000035.1 | NP_001000035.1 | 1 | 817 | ||
| Gene Symbol: Olr1514 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000036.1 | NP_001000036.1 | 1 | 817 | ||

Olr1513
Genomic Map