siRNA Details
| Gene Symbol: Dph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105809.2 | NP_001099279.1 | 7, 8 | 766 | ||
| GenBank mRNA | BC166808.1 | - | 7, 8 | 766 | 7124384 | |
| FQ228687.1 | - | 7, 8 | 782 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dph1| Gene Symbol: Dph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105809.2 | NP_001099279.1 | 7, 8 | 766 | ||
| GenBank mRNA | BC166808.1 | - | 7, 8 | 766 | 7124384 | |
| FQ228687.1 | - | 7, 8 | 782 | |||