siRNA Details
| Gene Symbol: Serpinf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177927.2 | NP_808788.1 | 7 | 950 | ||
| GenBank mRNA | AY216659.1 | - | 7 | 950 | ||
| BC078686.1 | - | 7 | 938 | 7110230 | ||
| FQ218413.1 | - | 7 | 951 | |||
| FQ219756.1 | - | 7 | 950 | |||
| FQ220670.1 | - | 1 | 85 | |||

Serpinf1
Genomic Map