siRNA Details
| Gene Symbol: Serpinf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011892.1 | NP_001011892.1 | 10 | 1148 | ||
| XM_006246675.1 | XP_006246737.1 | 10 | 1293 | |||
| GenBank mRNA | BC079362.1 | - | 10 | 1148 | 7109243 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Serpinf2| Gene Symbol: Serpinf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011892.1 | NP_001011892.1 | 10 | 1148 | ||
| XM_006246675.1 | XP_006246737.1 | 10 | 1293 | |||
| GenBank mRNA | BC079362.1 | - | 10 | 1148 | 7109243 | |