siRNA Details
| Gene Symbol: Slc43a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105812.1 | NP_001099282.1 | 11 | 1438 | ||
| XM_006246893.2 | XP_006246955.1 | 12 | 1728 | |||
| XM_006246894.2 | XP_006246956.1 | 12 | 1504 | |||
| XM_006246895.2 | XP_006246957.1 | 12 | 1477 | |||
| XM_006246896.2 | XP_006246958.1 | 12 | 1844 | |||
| XM_006246897.2 | XP_006246959.1 | 11 | 1713 | |||

Slc43a2
Genomic Map