siRNA Details
| Gene Symbol: Slc43a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105812.1 | NP_001099282.1 | 4 | 508 | ||
| XM_006246893.2 | XP_006246955.1 | 4 | 786 | |||
| XM_006246894.2 | XP_006246956.1 | 4 | 562 | |||
| XM_006246895.2 | XP_006246957.1 | 4 | 535 | |||
| XM_006246896.2 | XP_006246958.1 | 4 | 902 | |||
| XM_006246897.2 | XP_006246959.1 | 4 | 783 | |||

Slc43a2
Genomic Map